Learn More
Potassium acetate, 99%, Thermo Scientific Chemicals
Description
Potassium acetate is used as deicer to remove snow and ice from the surface and considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive, as a buffer in biological research, as an extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins, thereby removal of proteins from DNA. It plays an important role in preserving tissues, fixation and mummification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium acetate is used as deicer to remove snow and ice from the surface and considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive, as a buffer in biological research, as an extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins, thereby removal of proteins from DNA. It plays an important role in preserving tissues, fixation and mummification.
Solubility
Soluble in water, alcohol and ammonia. Insoluble in acetone and ether.
Notes
Hygroscopic. Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 292°C |
Density | 1.57 |
Quantity | 500 g |
Beilstein | 3595449 |
Sensitivity | Hygroscopic |
Merck Index | 14,7605 |
Solubility Information | Soluble in water,alcohol and ammonia. Insoluble in acetone and ether. |
Formula Weight | 98.15 |
Percent Purity | 99% |
Chemical Name or Material | Potassium acetate |
Safety and Handling
Hazard Category | H500 |
EINECSNumber | 204-822-2 |
RTECSNumber | AJ3325000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.