Potassium acetate, ACS, 99% min, Alfa Aesar™

Manufacturer: Alfa Aesar 13449F7

Quantity 5 x 2kg

 View more versions of this product

Catalog No. AA13449F7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127-08-2
    Physical Form Crystalline
    Assay Percent Range 99% min
    Molecular Formula C2H3KO2
    Formula Weight 98.15
    MDL Number MFCD00012458
    Chemical Name or Material Potassium acetate, For ACS analysis

    Specifications

    Formula Weight 98.15
    Quantity 5 x 2kg
    EINECS Number 204-822-2
    Physical Form Crystalline
    Density 1.57g/mL
    Assay Percent Range 99% min
    Molecular Formula C2H3KO2
    MDL Number MFCD00012458
    CAS 127-08-2
    Chemical Name or Material Potassium acetate, For ACS analysis

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit