Learn More
Potassium Acetate (Colorless Crystals or Powder), Fisher BioReagents
Description
Poly Bottle; 500g
Specifications
Potassium Acetate | |
White | |
7.0 to 8.0 | |
500 g | |
C2H3KO2 | |
15, 7726 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
Pass Test |
292°C | |
127-08-2 | |
Solid | |
≥99.0 % | |
CH3COOK | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
≥99.0% | |
Poly Bottle |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0
Recommended Storage : RT
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.