Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium benzofurazan-5-trifluoroborate, 95%

Click to view available options
Quantity:
1g
5g
1225608-24-1
C6H3BF3KN2O
226.007
MFCD09800741
VVXGHNBCIPLVLI-UHFFFAOYSA-N
potassium benzofurazan-5-trifluoroborate,potassium benzo c 1,2,5 oxadiazole-5-trifluoroborate,potassium 2,1,3-benzoxadiazol-5-yltrifluoroboranuide,amtb089,potassium 2,1,3-benzoxadiazol-5-yl trifluoroboranuide,potassium benzo c 1,2,5 oxadiazol-5-yltrifluoroborate,potassium 2,1,3-benzoxadiazol-5-yl trifluoro borate 1-
46738069
potassium;2,1,3-benzoxadiazol-5-yl(trifluoro)boranuide
[B-](C1=CC2=NON=C2C=C1)(F)(F)F.[K+]
1225608-24-1
C6H3BF3KN2O
226.007
MFCD09800741
VVXGHNBCIPLVLI-UHFFFAOYSA-N
potassium benzofurazan-5-trifluoroborate,potassium benzo c 1,2,5 oxadiazole-5-trifluoroborate,potassium 2,1,3-benzoxadiazol-5-yltrifluoroboranuide,amtb089,potassium 2,1,3-benzoxadiazol-5-yl trifluoroboranuide,potassium benzo c 1,2,5 oxadiazol-5-yltrifluoroborate,potassium 2,1,3-benzoxadiazol-5-yl trifluoro borate 1-
46738069
potassium;2,1,3-benzoxadiazol-5-yl(trifluoro)boranuide
[B-](C1=CC2=NON=C2C=C1)(F)(F)F.[K+]
Quantity 1g
Formula Weight 226.01
Percent Purity 95%
Chemical Name or Material Potassium benzofurazan-5-trifluoroborate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures