Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bicarbonate, Granular, FCC, 99-101.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
298-14-6
CHKO3
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium hydrogen carbonate
[K+].OC([O-])=O
298-14-6
CHKO3
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium hydrogen carbonate
[K+].OC([O-])=O

P1182, 298-14-6, KHCO3

Spectrum™ Potassium Bicarbonate, Granular, FCC is used as a carbon dioxide source for leavening in baking. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 99 to 101.5%
Lead (Pb) 2 mg/kg
Loss on Drying 0.0025
Packaging Poly Pail
Quantity 12 kg