Alfa Aesar™ Potassium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.997% (metals basis)

Manufacturer: Alfa Aesar™ 4289809

Quantity 10g

 View more versions of this product

Catalog No. AA4289809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14402-67-6
    Molecular Formula C4K2O9Ti·xH2O
    Linear Formula K2[TiO(C2O4)2]·xH2O
    Formula Weight 318.10 (Anhydrous)
    MDL Number MFCD00150592
    UN Number UN3288
    Synonym Potassium titanyloxolate
    Chemical Name or Material Potassium bis (oxalato)oxotitanate (IV) hydrate
    Grade Puratronic™

    Specifications

    Physical Form Coarse Crystalline Powder
    Formula Weight 318.10 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00150592
    UN Number UN3288
    Synonym Potassium titanyloxolate
    Health Hazard 1 Warning
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Molecular Formula C4K2O9Ti·xH2O
    Linear Formula K2[TiO(C2O4)2]·xH2O
    CAS 14402-67-6
    Odor Odorless
    Color Colorless
    Quantity 10g
    Chemical Name or Material Potassium bis (oxalato)oxotitanate (IV) hydrate
    Assay 99.997%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit