Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 4289818

Quantity 50g

 View more versions of this product

Catalog No. AA4289818


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14402-67-6
K2[TiO(C2O4)2]·xH2O
MFCD00150592
Potassium titanyloxolate
Puratronic™
C4K2O9Ti·xH2O
318.10 (Anhydrous)
UN3288
Potassium bis (oxalato)oxotitanate (IV) hydrate

Specifications

14402-67-6
C4K2O9Ti·xH2O
318.10 (Anhydrous)
UN3288
Odorless
50g
99.997%, (metals basis)
Coarse Crystalline Powder
K2[TiO(C2O4)2]·xH2O
MFCD00150592
Potassium titanyloxolate
Colorless
Potassium bis (oxalato)oxotitanate (IV) hydrate
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit