Alfa Aesar™ Potassium bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 4690903

Quantity 1g

 View more versions of this product

Catalog No. AA4690903

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 90076-67-8
    Molecular Formula C2F6KNO4S2
    Linear Formula (CF3SO2)2NK
    Formula Weight 319.24
    MDL Number MFCD06200829
    UN Number UN1759
    Synonym Potassium triflimide; Potassium trifluoromethanesulfonimide
    Chemical Name or Material Potassium bis (trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 319.24
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    Health Hazard 1 Danger
    Linear Formula (CF3SO2)2NK
    MDL Number MFCD06200829
    CAS 90076-67-8
    UN Number UN1759
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point 198° to 203°C
    Sensitivity Moisture Sensitive
    Molecular Formula C2F6KNO4S2
    Synonym Potassium triflimide; Potassium trifluoromethanesulfonimide
    Quantity 1g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Chemical Name or Material Potassium bis (trifluoromethylsulfonyl)imide

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit