Potassium bis(trimethylsilyl)amide, 15% w/w in toluene, Alfa Aesar™

Manufacturer: Alfa Aesar 44143AP

Quantity 500mL

 View more versions of this product

Catalog No. AA44143AP

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 40949-94-8
    Physical Form Liquid
    Molecular Formula C6H18KNSi2
    Formula Weight 199.49
    MDL Number MFCD00010330
    Synonym Potassium bis(trimethylsilyl)amide
    Chemical Name or Material Potassium bis(trimethylsilyl)amide

    Specifications

    Formula Weight 199.49
    Quantity 500mL
    EINECS Number 424-100-2
    Physical Form Liquid
    Density 0.881g/mL
    MDL Number MFCD00010330
    CAS 40949-94-8
    Synonym Potassium bis(trimethylsilyl)amide
    Molecular Formula C6H18KNSi2
    Chemical Name or Material Potassium bis(trimethylsilyl)amide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit