Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bis(trimethylsilyl)amide, 15% w/w in toluene, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™

Manufacturer:  Alfa Aesar 44143AC

Packaging Packaged under Argon in resealable ChemSeal™ bottles
Quantity 25mL

 View more versions of this product

Catalog No. AA44143AC

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

40949-94-8
[(CH3)3Si]2NK
MFCD00010330
4006754
Potassium bis(trimethylsilyl)amide
C6H18KNSi2
199.49
UN2924
Potassium hexamethyldisilazide

Specifications

40949-94-8
C6H18KNSi2
199.49
UN2924
Potassium hexamethyldisilazide
1.4920
Colorless to Amber
25mL
Air and Moisture Sensitive
Liquid
[(CH3)3Si]2NK
MFCD00010330
4006754
Packaged under Argon in resealable ChemSeal™ bottles
0.881
15% w/w in toluene
7°C (44°F)
Potassium bis(trimethylsilyl)amide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H361-H373-H336;
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P301+P310-P305+P351+P338-P315-P402;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Get immediate medical advice/attention.
Store in a dry place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit