Alfa Aesar™ Potassium bis(trimethylsilyl)amide, 20% w/w, 0.91M in THF

Manufacturer: Alfa Aesar™ 35450AK

Quantity 250mL

 View more versions of this product

Catalog No. AA35450AK

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 40949-94-8
    Linear Formula [(CH3)3Si]2NK
    Formula Weight 199.49
    MDL Number MFCD00010330
    UN Number UN2924
    Synonym Potassium hexamethyldisilazide
    Chemical Name or Material Potassium bis(trimethylsilyl)amide

    Specifications

    Formula Weight 199.49
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00010330
    UN Number UN2924
    Synonym Potassium hexamethyldisilazide
    Health Hazard 1 Danger
    Density 0.910g/mL
    Boiling Point 66°C
    Color Colorless to amber
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Linear Formula [(CH3)3Si]2NK
    CAS 40949-94-8
    Health Hazard 2 GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.
    Concentration 20% w/w, 0.91M in THF
    Quantity 250mL
    Chemical Name or Material Potassium bis(trimethylsilyl)amide

    Description

    • Used in the preparation of lanthanide complexes, selective cyclization reactions, and as an anionic initiator for polymerization

    GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit