Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium bis(trimethylsilyl)amide, 20% w/w, 0.91M in THF

Manufacturer:  Alfa Aesar™ 35450K2

Quantity 1L

 View more versions of this product

Catalog No. AA35450K2

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Used in the preparation of lanthanide complexes, selective cyclization reactions, and as an anionic initiator for polymerization
Description & Specifications

Chemical Identifiers

40949-94-8
199.49
UN2924
Potassium bis(trimethylsilyl)amide
[(CH3)3Si]2NK
MFCD00010330
Potassium hexamethyldisilazide

Specifications

40949-94-8
[(CH3)3Si]2NK
MFCD00010330
Potassium hexamethyldisilazide
66°C
20% w/w, 0.91M in THF
Air & Moisture Sensitive
Liquid
199.49
UN2924
0.910g/mL
Colorless to amber
1L
Potassium bis(trimethylsilyl)amide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit