Alfa Aesar™ Potassium bis(trimethylsilyl)amide, 20% w/w, 0.91M in THF, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 44208K2

Quantity 1L

 View more versions of this product

Catalog No. AA44208K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 40949-94-8
    Molecular Formula C6H18KNSi2
    Linear Formula [(CH3)3Si]2NK
    Formula Weight 199.49
    MDL Number MFCD00010330
    UN Number UN2924
    Synonym Potassium hexamethyldisilazide
    Chemical Name or Material Potassium bis (trimethylsilyl)amide

    Specifications

    Formula Weight 199.49
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H18KNSi2
    Synonym Potassium hexamethyldisilazide
    Sensitivity Air & Moisture Sensitive
    CAS 40949-94-8
    Health Hazard 2 GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.
    Color Colorless to amber
    Flash Point −14°C (6°F)
    Physical Form Liquid
    UN Number UN2924
    Packaging ChemSeal™ bottles
    Concentration 0.91M in THF
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Linear Formula [(CH3)3Si]2NK
    MDL Number MFCD00010330
    Density 0.910g/mL
    Boiling Point 66°C
    Storage Note 1
    • Packaged under Argon
    Health Hazard 1 Danger
    Quantity 1L
    Chemical Name or Material Potassium bis (trimethylsilyl)amide
    Assay 20% (w/w)

    GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit