Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium-Bismuth Iodide TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle

P-213, 41944-01-8

Spectrum™ Potassium-Bismuth Iodide TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS 7681-11-0,87-69-4
CAS Max % 0.001,0.999
Molecular Formula IK
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium iodide
Molecular Weight (g/mol) 166.00
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL