Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bisulfate, 99%, extra pure, Acros Organics

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
7646-93-7
HKO4S
136.16
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate,potassium hydrogen sulfate,potassium bisulphate,sulfuric acid, monopotassium salt,potassium acid sulfate,sal enixum,potassium hydrogensulphate,monopotassium sulfate,acid potassium sulfate,caswell no. 682c
516920
potassium;hydrogen sulfate
OS(=O)(=O)[O-].[K+]
7646-93-7
HKO4S
136.16
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate,potassium hydrogen sulfate,potassium bisulphate,sulfuric acid, monopotassium salt,potassium acid sulfate,sal enixum,potassium hydrogensulphate,monopotassium sulfate,acid potassium sulfate,caswell no. 682c
516920
potassium;hydrogen sulfate
OS(=O)(=O)[O-].[K+]
Assay Percent Range 99%
Linear Formula KHSO4
Ammonium 0.003% max
Merck Index 15,7734
Solubility Information Solubility in water: 490g/L (20°C). Other solubilities: decomposes in alcohol
Formula Weight 136.16
Percent Purity 99%
Grade Extra Pure
Chloride 0.002% max
Packaging Plastic drum
Solubility (10% in water) Clear colorless
Melting Point 214.0°C
Quantity 10kg
Chemical Name or Material Potassium bisulfate
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN

GHS Signal Word: Danger

EINECSNumber : 231-594-1