Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bisulfate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7646-93-7
HKO4S
136.162
CHKVPAROMQMJNQ-UHFFFAOYSA-M
516920

High quality chemicals for laboratory and specialized industrial use

SMILES OS(=O)(=O)[O-].[K+]
IUPAC Name potassium;hydrogen sulfate
Formula Weight 136.17
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.24kg
Quantity 2.5kg
Chemical Name or Material Potassium Bisulfate
Grade BAKER ANALYZED™ Reagent