Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bisulfate, Reagent Grade, Reagents

Catalog No. 01337872
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-872 500 g
01-337-873 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-872 Supplier Reagents Holdings Llc Supplier No. C1277500500A5
Only null left
Add to Cart
Add to Cart

Potassium Bisulfate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym Potassium Hydrogen Sulfate
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Boiling Point 330°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.32
Chemical Name or Material Potassium Bisulfate, Reagent Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.