Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bitartrate, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
868-14-4
C4H5KO6
188.18
KYKNRZGSIGMXFH-ZVGUSBNCSA-M
potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
868-14-4
C4H5KO6
188.18
KYKNRZGSIGMXFH-ZVGUSBNCSA-M
potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

P1801, 868-14-4, C4H5KO6

Spectrum™ Potassium Bitartrate, USP is used internally as a purgative. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 99 to 101%
Packaging Poly Pail
Quantity 2.5 kg