Alfa Aesar™ Potassium borohydride, 98%

Manufacturer: Alfa Aesar™ 4684722

Quantity 100g

 View more versions of this product

Catalog No. AA4684722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13762-51-1
    Molecular Formula BH4K
    Linear Formula KBH4
    Formula Weight 53.94
    MDL Number MFCD00011396
    UN Number UN1870
    Merck Index 14,7616
    Synonym Potassium tetrahydridoborate
    Chemical Name or Material Potassium borohydride

    Specifications

    Formula Weight 53.94
    Health Hazard 2 GHS H Statement
    H261-H301-H311-H331-H314-H318
    In contact with water releases flammable gas.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Linear Formula KBH4
    Quantity 100g
    Health Hazard 1 Danger
    Density 1.17g/mL
    Refractive Index 1.494
    Molecular Formula BH4K
    UN Number UN1870
    Synonym Potassium tetrahydridoborate
    Melting Point >400°C (dec.)
    Solubility Information Soluble in water (19g/100g ); methanol (3.9g/100g ); 4:1 water:methanol mixture (13g/100g );
    Physical Form Granules
    Sensitivity Moisture Sensitive
    CAS 13762-51-1
    Merck Index 14,7616
    TSCA TSCA
    Mesh Size −4 Mesh
    Odor Odorless
    MDL Number MFCD00011396
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: POTASSIUM BOROHYDRIDE
    RTECS Number TS7525000
    Chemical Name or Material Potassium borohydride
    Assay 98%

    Description

    • Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics

    GHS H Statement
    H261-H301-H311-H331-H314-H318
    In contact with water releases flammable gas.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit