Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium borohydride, 98+%, Acros Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
13762-51-1
H4BK
53.93
MFCD00011396
HHUJLKPGQMFFMS-UHFFFAOYSA-N
potassium borohydride,potassium tetrahydroborate,potassium borohydrate,unii-0ws230dtgf,potassium tetrahydroborate 1-,borohydrure de potassium french,borate 1-, tetrahydro-, potassium,0ws230dtgf,borohydrure de potassium,borylpotassium
22892188
potassium;boron(1-)
[B-].[K+]
13762-51-1
H4BK
53.93
MFCD00011396
HHUJLKPGQMFFMS-UHFFFAOYSA-N
potassium borohydride,potassium tetrahydroborate,potassium borohydrate,unii-0ws230dtgf,potassium tetrahydroborate 1-,borohydrure de potassium french,borate 1-, tetrahydro-, potassium,0ws230dtgf,borohydrure de potassium,borylpotassium
22892188
potassium;boron(1-)
[B-].[K+]
Reduction reagent
Assay Percent Range 98+%
Linear Formula KBH4
Fieser 04,398; 06,476; 08,407
Merck Index 15, 7737
Solubility Information Solubility in water: 190g/L (25°C). Other solubilities: reacts with water,partially soluble in ammonia,insoluble in ethers,hydrocarbons,isopropylamine,,benzene,hexane,dioxane,thf and acetonitrile,7g/L methanol (20°C)
Formula Weight 53.93
Physical Form Crystalline Powder
Percent Purity 98+%
Packaging Glass bottle
Sodium (Na) 1% max.
Solubility Solubility in water: 190g/L (25°C). Other solubilities: reacts with water, partially soluble in ammonia, insoluble in ethers, hydrocarbons, isopropylamine,, benzene, hexane, dioxane, thf and acetonitrile, 7g/L methanol (20°C)
Color White
Melting Point 500.0°C
Quantity 5g
Chemical Name or Material Potassium borohydride, 98%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
In contact with water releases flammable gas.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 237-360-5

RTECSNumber : TS7525000

TSCA : TSCA