Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromate, 0.0913 Normal Standardized,, Reagents

Catalog No. 01334373
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-373 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-373 Supplier Reagents Holdings Llc Supplier No. CP1117001C

Due to product restrictions, please to purchase or view availability for this product.

Potassium Bromate, 0.0913 Normal Standardized, NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Potassium Bromate, 0.0913 Normal Standardized, NIST Traceable
Boiling Point 100°C
Color Colorless
CAS 7732-18-5
Physical Form Liquid
Quantity 1 L
Molecular Formula KBrO3/H2O
Synonym Bromic Acid Potassium Salt
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0018
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.