Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromate, 99+%, extra pure, Acros Organics

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
7758-01-2
BrKO3
167.01
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate,bromic acid, potassium salt,bromate, potassium,potassium bromate dot,unii-04mb35w6za,ccris 529,eec no. e924,bromic acid, potassium salt 1:1,potassiumbromate,kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]
7758-01-2
BrKO3
167.01
MFCD00011359
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium bromate,bromic acid, potassium salt,bromate, potassium,potassium bromate dot,unii-04mb35w6za,ccris 529,eec no. e924,bromic acid, potassium salt 1:1,potassiumbromate,kaliumbromat
23673461
CHEBI:38211
potassium;bromate
[O-]Br(=O)=O.[K+]
C-X bond formation reagent, Oxidation reagent
Assay Percent Range 99% min. (Iodometry)
Linear Formula KBrO3
Fieser 11,431
Merck Index 15, 7738
Solubility Information Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in boiling water
Formula Weight 167.01
Physical Form Liquid
Percent Purity 99+%
Grade Extra Pure
Bromide 500ppm max.
Bromine (Br) 500ppm max.
Density 3.2700g/mL
Heavy Metals (as Pb) 20ppm max.
Lead (Pb) 5ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water
Specific Gravity 3.27
Sulfate 100ppm max.
Color Colorless
Melting Point 350.0°C
Quantity 2.5kg
Chemical Name or Material Potassium bromate, 99+%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
May cause cancer.
May cause fire or explosion; strong oxidizer.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-829-8

RTECSNumber : EF8725000

TSCA : TSCA