Learn More
Potassium bromate, 99+%, extra pure, ACROS Organics™
Description
Specifications
Assay Percent Range | 99% min. (Iodometry) |
Linear Formula | KBrO3 |
Fieser | 11,431 |
Merck Index | 15, 7738 |
Solubility Information | Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in boiling water |
Formula Weight | 167.01 |
Physical Form | Liquid |
Percent Purity | 99+% |
Grade | Extra Pure |
Bromide | 500ppm max. |
Show More |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause cancer.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-829-8
RTECSNumber : EF8725000
TSCA : TSCA