Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, 98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
7758-02-3
BrK
119
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
7758-02-3
BrK
119
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
Alkaline Earth Metals 0.05% max. (as Ba)
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Linear Formula KBr
Merck Index 15, 7739
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol
Formula Weight 119
Physical Form Crystalline Powder and Chunks
Percent Purity 98%
Grade Extra Pure
Chloride 4500ppm max
Chlorine (Cl) 4500ppm max.
Heavy Metals (as Pb) 10ppm max.
Iodine (I) 200ppm max.
Iron (Fe) 10ppm max.
Loss on Drying 1% max.
Oxidizing Substances 0.001% max. (as KBrO3)
Packaging Plastic bottle
Solubility Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
Sulfate 50ppm max.
Boiling Point 1435.0°C
Color White
Melting Point 734.0°C
Quantity 1kg
Chemical Name or Material Potassium bromide, Extra Pure
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 231-830-3