Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, 99.92% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
25g
100g
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
Assay Percent Range 99.92% (metals basis)
Appearance White Powder/crystalline/beads
Solubility Information Freely soluble in water. Soluble in alcohol and glycerol. Slightly soluble in diethyl ether.
Formula Weight 119.01
Physical Form Crystalline
Density 2.75
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Boiling Point 1435°C
Color Colorless
Melting Point 730°C
Quantity 25g
Chemical Name or Material Potassium bromide
Show More Show Less

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

EINECSNumber : 231-830-3

RTECSNumber : TS7650000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen