Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, 99+%, for spectroscopy, IR grade, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
7758-02-3
BrK
119
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
7758-02-3
BrK
119
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide,bromide salt of potassium,kalii bromidum,bromure de potassium,caswell no. 684,tripotassium tribromide,potassium bromide k3br3,unii-osd78555zm,ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
For IR spectroscopy
Absorbance water: 3250-3750/1620-1640 1/cm: < 0.15, CH-compounds: 2750-3100 1/cm: < 0.004, other: 420-4000 1/cm: < 0.015
Absorbance Conditions 2g pressed disk, 0.5cm layer thickness
Additional Information application test: passes
Assay Percent Range 99% min. (Argentometry)
Linear Formula KBr
Merck Index 15, 7739
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol
Formula Weight 119
Physical Form Fine Crystalline Powder
Percent Purity 99+%
Grade Spectroscopy
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
Boiling Point 1435.0°C
Color White
Melting Point 734.0°C
Quantity 500g
Chemical Name or Material Potassium bromide, 99+%
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 231-830-3

RTECSNumber : TS7650000

TSCA : TSCA