Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
100g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7758-02-3
BrK
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    For Infrared Analysis
Formula Weight 118.99
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.75kg
Quantity 100g
Chemical Name or Material Potassium Bromide
Grade BAKER INSTRA-ANALYZED™ Reagent