Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, Crystal, IR Grade, Spectrum™

Click to view available options
Quantity:
25 g
100 g
6 x 25 g
Packaging:
Amber Glass Bottle
7758-02-3
BrK
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
[K+].[Br-]
7758-02-3
BrK
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
[K+].[Br-]

P1225, 7758-02-3, KBr

Spectrum™ Potassium Bromide, Crystal, IR Grade is used widely as infrared optical windows and components for general spectroscopy because it is transparent from the near ultraviolet to long-wave infrared wavelengths and has no significant optical absorption lines in its high transmission region.

Grade IR Grade
CAS Max % 1
Calcium (Ca) 2e-005
Heavy Metals (as Pb) 5e-006
Insoluble Matter 5e-005
Iron (Fe) 5e-006
Magnesium (Mg) 1e-005
Packaging Amber Glass Bottle
Sodium (Na) 0.0002
pH 5.0 to 8.8
Quantity 25 g
Show More Show Less