Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Bromide, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
7758-02-3
BrK
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
[K+].[Br-]
7758-02-3
BrK
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
[K+].[Br-]

P1742, 7758-02-3, KBr

Spectrum™ Potassium Bromide, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 98 to 100.5%
Iron (Fe) 20 ppm
Loss on Drying 0.01
Packaging Amber Glass Bottle
Sulfate 0.0001
Quantity 2.5 kg