Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromomethyltrifluoroborate, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
888711-44-2
CH2BBrF3K
200.83
AZDFPIRYUOCVCJ-UHFFFAOYSA-N
potassium bromomethyl trifluoroborate,potassium bromomethyltrifluoroborate,potassium bromomethyl trifluoroboranuide,potassium bromomethyl trifluoro boranuide,potassium bromomethyl trifluoro borate 1-,borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4,pubchem11564,bromomethyl potassium trifluoroborate,postassium bromomethyltrifluoroborate,trifluoro bromomethyl potassioboron v
23690312
potassium;bromomethyl(trifluoro)boranuide
[B-](CBr)(F)(F)F.[K+]
888711-44-2
CH2BBrF3K
200.83
AZDFPIRYUOCVCJ-UHFFFAOYSA-N
potassium bromomethyl trifluoroborate,potassium bromomethyltrifluoroborate,potassium bromomethyl trifluoroboranuide,potassium bromomethyl trifluoro boranuide,potassium bromomethyl trifluoro borate 1-,borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4,pubchem11564,bromomethyl potassium trifluoroborate,postassium bromomethyltrifluoroborate,trifluoro bromomethyl potassioboron v
23690312
potassium;bromomethyl(trifluoro)boranuide
[B-](CBr)(F)(F)F.[K+]
Assay Percent Range 95%
Formula Weight 200.83
Physical Form Powder
Percent Purity 95%
Elemental Analysis 5.62 to 6.33% C, 0.94 to 1.06%
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 220.0°C
Quantity 1g
Chemical Name or Material Potassium bromomethyltrifluoroborate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning