Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate (15% w/v), Ricca Chemical

Click to view available options
Quantity:
500mL
584-08-7,7732-18-5
CK2O3
138.205
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
584-08-7,7732-18-5
CK2O3
138.205
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
CAS Min % 85.0,12.79
CAS Max % 89.0,13.31
Quantity 500mL
Chemical Name or Material Potassium Carbonate (15% w/v)