Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium carbonate, 98%, extra pure, anhydrous, Acros Organics

Click to view available options
Quantity:
1kg
5kg
25kg
584-08-7
CK2O3
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
584-08-7
CK2O3
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
Linear Formula K2CO3
Merck Index 15,774
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
Formula Weight 138.21
Percent Purity 98%
Grade Extra Pure
Heavy Metals (as Pb) 0.005% max.
Loss on Drying 2.0% max.
Solubility Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
Melting Point 891.0°C
Quantity 25kg
Chemical Name or Material Potassium carbonate
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri

GHS Signal Word: Warning

EINECSNumber : 209-529-3