Potassium carbonate, ACS, 99.0% min, Alfa Aesar™

Manufacturer: Alfa Aesar 12609A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12609A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 584-08-7
    Molecular Formula K2CO3
    Formula Weight 138.21
    MDL Number MFCD00011382
    Beilstein 4267587
    Merck Index 14,7619
    Chemical Name or Material Potassium carbonate
    Grade ACS Grade

    Specifications

    Formula Weight 138.21
    Quantity 1kg
    TSCA TSCA
    Physical Form Granular
    Sensitivity Hygroscopic
    MDL Number MFCD00011382
    Merck Index 14,7619
    Density 2.29
    RTECS Number TS7750000
    Molecular Formula K2CO3
    CAS 584-08-7
    Beilstein 4267587
    Melting Point 891°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335;
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Potassium carbonate
    Assay ≥99.0%
    Grade ACS Grade

    GHS H Statement
    H302-H315-H319-H335;
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit