Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, ACS Reagent, ≥99.0%, Honeywell™ Fluka™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
Packaging:
Poly Bottle
584-08-7
CK2O3
138.205
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
584-08-7
CK2O3
138.205
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
Appearance Powder
Formula Weight 138.21g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Bulk Density ca. 1000kg/cu.m
Density 2.430g/cu.cm
Packaging Poly Bottle
Physical State Solid
Color White
Melting Point 891°C
pH 11.6 (at 20°C)
Quantity 500g
Chemical Name or Material Potassium Carbonate
Show More Show Less

P261-P305 + P351 + P338

H302-H315-H319-H335

EINECSNumber : 209-529-3

RTECSNumber : TS7750000