Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, Anhydrous, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

584-08-7
CK2O3
138.205
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430

High quality chemicals for laboratory and specialized industrial use

CAS 584-08-7
Molecular Formula CK2O3
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;carbonate
Molecular Weight (g/mol) 138.205
PubChem CID 11430
Formula Weight 138.21
Physical Form Granular
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.29kg
Quantity 500g
Chemical Name or Material Potassium Carbonate, Anhydrous
Grade Baker Analyzed™ ACS Reagent
Show More Show Less