Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Carbonate, Anhydrous, Reagents
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
Specifications
Specifications
Color | Colorless/White |
pH | 11.6 |
Physical Form | Solid |
Quantity | 50 lb. |
Grade | Reagent |
Packaging | Fiber Drum |
Specific Gravity | 2.29 |
Chemical Name or Material | Potassium Carbonate, Anhydrous |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction