Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Potassium carbonate BioXtra, Honeywell Fluka™

Click to view available options
Quantity:
500g
1kg
584-08-7
CK2O3
138.205
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
584-08-7
CK2O3
138.205
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]

BioXtra,≥99.0%

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
Formula Weight 138.21g/mol
Percent Purity ≥99%
Quantity 1kg
Chemical Name or Material Potassium carbonate

P280-P301 + P312 + P330-P305 + P351 + P338-P337 + P313

RTECSNumber : TS7750000