Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Potassium carbonate, purum p.a., Honeywell

Click to view available options
Quantity:
1kg
5kg
584-08-7
CK2O3
138.205
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]
584-08-7
CK2O3
138.205
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate,dipotassium carbonate,potash,potassium carbonate, anhydrous,carbonic acid, dipotassium salt,carbonate of potash,pearl ash,salt of tartar,kaliumcarbonat,potassiumcarbonate
11430
dipotassium;carbonate
C(=O)([O-])[O-].[K+].[K+]

Purum p.a., anhydrous,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
Formula Weight 138.21g/mol
Percent Purity ≥99% (T)
Grade purum p.a.
Quantity 5kg
Chemical Name or Material Potassium carbonate

P261-P305 + P351 + P338

RTECSNumber : TS7750000