Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium carbonate sesquihydrate, ACS, 98.5-101.0%

Manufacturer:  Alfa Aesar 1098136

Quantity 500g

 View more versions of this product

Catalog No. AA1098136


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6381-79-9
165.23 (138.21anhydrous)
14,7670
ACS Grade
K2CO3·1·5H2O
MFCD00149925
Potassium carbonate sesquihydrate

Specifications

6381-79-9
98.5-101.0%
White crystals
MFCD00149925
Soluble in water; Practically insoluble in alcohol
Odorless
Potassium carbonate sesquihydrate
ACS Grade
Crystalline
K2CO3·1·5H2O
165.23 (138.21anhydrous)
14,7670
Maximum level of impurities: Insoluble matter 0.01%, Cl 0.003%, PO4
500g
98.5 to 101.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit