Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Carbonate, Sesquihydrate, Crystal, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6381-79-9
C2H6K4O9
330.45
WWVGGBSGYNQKCY-UHFFFAOYSA-J
tetrapotassium trihydrate dicarbonate
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
6381-79-9
C2H6K4O9
330.45
WWVGGBSGYNQKCY-UHFFFAOYSA-J
tetrapotassium trihydrate dicarbonate
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

PO162, 6381-79-9, K2CO3•1.5H2O

Spectrum™ Potassium Carbonate, Sesquihydrate, Crystal, FCC is used as a meat tenderizer, a source of dietary potassium and a buffering agent in wine production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 99.0 to 100.5%
Lead (Pb) 2 mg/kg
Loss on Drying 10-16.5%
Packaging Poly Pail
Quantity 2.5 kg