Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium chlorate, ACS, 99.0% min, Alfa Aesar™
Click to view available options
Quantity:
500g
2kg
Specifications
Assay Percent Range | 99.0% |
Solubility Information | Soluble in water,glycerol. Insoluble in alcohol. |
Formula Weight | 122.55 |
Physical Form | Crystalline |
Grade | ACS Reagent |
Density | 2.32 g/mL |
Sensitivity | Hygroscopic |
Boiling Point | 400°C (decomposition) |
Melting Point | 368°C |
Quantity | 500g |
Show More |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
H271-H302+H332
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 223-289-7
RTECSNumber : FO0350000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen