Alfa Aesar™ Potassium chloride, 0.005M, Conductivity Standard

Manufacturer: Alfa Aesar™ 42680AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42680AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7
    Formula Weight 74.55
    MDL Number MFCD00011360
    Chemical Name or Material Potassium chloride Conductivity Standard, 718μmho at 25°C

    Specifications

    Formula Weight 74.55
    Quantity 500mL
    MDL Number MFCD00011360
    TSCA TSCA
    Physical Form Liquid
    Concentration 0.005M
    CAS 7447-40-7
    Odor Odorless
    Chemical Name or Material Potassium chloride Conductivity Standard, 718μmho at 25°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit