Potassium chloride, 0.117M, Conductivity Standard, Alfa Aesar™

Manufacturer: Alfa Aesar 42696K7

Quantity 4L

 View more versions of this product

Catalog No. AA42696K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7,7732-18-5
    Physical Form Liquid
    Assay Percent Range 0.117M
    Molecular Formula KCl
    Formula Weight 74.55
    MDL Number MFCD00011360
    Chemical Name or Material Potassium chloride, 15, 000μmho at 25°C

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Formula Weight 74.55
    Quantity 4L
    EINECS Number 231-211-8
    Assay Percent Range 0.117M
    Appearance Colorless liquid
    Molecular Formula KCl
    MDL Number MFCD00011360
    CAS 7447-40-7,7732-18-5
    Concentration or Composition (by Analyte or Components) Potassium chloride: 0.88%; Water: 99.12%
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Potassium chloride, 15, 000μmho at 25°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit