Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, 0.85M, Conductivity Standard, Alfa Aesar™

Manufacturer:  Alfa Aesar 43405K7

Quantity 4L

 View more versions of this product

Catalog No. AA43405K7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7447-40-7,7732-18-5
0.85M
74.55
Potassium chloride
Liquid
KCl
MFCD00011360

Specifications

7447-40-7,7732-18-5
0.85M
74.55
Potassium chloride: 6.35%; Water: 93.65%
17mm Hg at 20°C
4L
Liquid
KCl
MFCD00011360
Odorless
Clear colorless
Potassium chloride

Safety and Handling

SDS & Safety Documents

EINECSNumber : 231-211-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit