Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride, 3 Molar Electrode Filling Solution, Reagents

Catalog No. p-200021368
Click to view available options
Quantity:
120 mL
250 mL
500 mL
1 L
4 L
10 L

Potassium Chloride, 3 Molar Electrode Filling Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Chloride, 3 Molar Electrode Filling Solution
Boiling Point 103°C
CAS 7732-18-5, 7447-40-7
Color Colorless
pH 5.4 to 8.6
Physical Form Liquid
Quantity 10 L
Molecular Formula KCl/H2O
Synonym Muriate of Potash
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.1129
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.