Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
7447-40-7
ClK
74.54
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
7447-40-7
ClK
74.54
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
Biological buffer for analysis
Acidity Alkalinity complies
Aluminum (Al) 1ppm max.
Assay Percent Range 99.0 to 100.5% (on dried substance) (Argentometry)
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Arsenic (As) 0.5ppm max.
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in ether and acetone
Formula Weight 74.54
Physical Form Liquid
Percent Purity 99+%
Grade Analysis
Bromide 1000ppm max.
Bromine (Br) 1000ppm max.
Cadmium (Cd) 1ppm max.
Calcium (Ca) 50ppm max.
Copper (Cu) 10ppm max.
Free Alkali 0.005% max.
Free Acid 0.003% max.
Heavy Metals (as Pb) 5ppm max.
Iodide 50ppm max.
Iron (Fe) 10ppm max.
Lead (Pb) 2ppm max.
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Magnesium (Mg) 50ppm max.
Packaging Plastic bottle
Sodium (Na) 700ppm max.
Solubility Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
Sulfate 100ppm max.
Trace Analysis 20ppm max.
Sublimation Point 1500.0°C
Boiling Point 1420.0°C
Color Colorless
Melting Point 770.0°C
Quantity 1kg
Chemical Name or Material Potassium chloride, p.a.
Show More Show Less

HYGROSCOPIC

EINECSNumber : 231-211-8

RTECSNumber : TS8050000

TSCA : TSCA