Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium chloride, 99%, Molecular Biology Grade

Click to view available options
Quantity:
500g
1kg
7447-40-7
ClK
74.548
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
7447-40-7
ClK
74.548
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride,enseal,muriate of potash,klotrix,sylvite,slow-k,potassium chloride kcl,klor-con,chlorvescent,kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
Merck Index 14,7621
Formula Weight 74.55
Physical Form Powder
Percent Purity 99%
Grade Molecular Biology
Density 1.984g/mL
Odor Odorless
Boiling Point 1500°C (sublimation)
Color White
Melting Point 773°C
Quantity 500g
Chemical Name or Material Potassium chloride
Show More Show Less

RTECSNumber : TS8050000

TSCA : TSCA