Potassium chloride, ACS, 99.0-100.5%

In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy

Manufacturer: Alfa Aesar 11595A7

Quantity 5kg

 View more versions of this product

Catalog No. AA11595A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7447-40-7
    Molecular Formula KCl
    Formula Weight 74.55
    MDL Number MFCD00011360
    Merck Index 14,7621
    Chemical Name or Material Potassium chloride, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 74.55
    Quantity 5kg
    Boiling Point 1500°C (subl.)
    Melting Point 773°C
    TSCA TSCA
    Merck Index 14,7621
    Solubility Information Freely soluble in water; Soluble in glycerol, alcohol
    RTECS Number TS8050000
    Physical Form Crystalline
    Density 1.984g/mL
    Assay Percent Range 99.0 to 100.5%
    MDL Number MFCD00011360
    CAS 7447-40-7
    Molecular Formula KCl
    Chemical Name or Material Potassium chloride, For analysis ACS
    Assay 99.0 to 100.5%
    Grade ACS Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit