Learn More
Potassium chloride, ACS, 99.0-100.5%
Description
Potassium chloride is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins. It acts as a fire extinguishing agent in portable and wheeled fire extinguishers. It may be used as a softener unit in household water, which is an alternative to sodium chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy

Specifications
Specifications
Boiling Point | 1500°C (sublimation) |
Melting Point | 773°C |
Physical Form | Crystalline |
Quantity | 250 g |
Assay Percent Range | 99.0 to 100.5% |
Merck Index | 14,7621 |
Solubility Information | Soluble in water,glycerol and alkalies; Slightly soluble in alcohol and insoluble in ether. |
IUPAC Name | potassium chloride |
Formula Weight | 74.55 |
Percent Purity | 99.0 to 100.5% |
Show More |
Safety and Handling
EINECSNumber | 231-211-8 |
RTECSNumber | TS8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.