Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chloride, ACS Reagent, Honeywell Fluka™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
potassium chloride
[Cl-].[K+]
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
potassium chloride
[Cl-].[K+]

ACS Reagent, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Quantity 4 x 2.5 kg
Linear Formula KCl
UN Number NONH for all modes of transport
Formula Weight 74.55g/mol
Percent Purity 99 to 100.5%
Grade ACS Reagent
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium chloride

RTECSNumber : TS8050000